Learn More
Labconco™ XVS™ PVC Elbow for Ventilation Stations
PVC Elbow for Ventilation Stations
Brand:  Labconco™ 4708700
Product Code. 15459444
  • £588.97 / Each
Explore more special offers
Description
Description
  • For use with Labconco Protector XVS Ventilation Stations
  • 90° elbow has belled end connections to receive PVC duct directly
Specifications
Specifications
Protector VS Ventilation Station
8 lb.
34.59 cm
6 in. dia. Round PVC Duct
Designed for quick installation and minimum pressure losses
13.62 in.
3.6 Kg
90° Elbow
PVC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ XVS™ PVC Elbow for Ventilation Stations > PVC Elbow