Learn More
Labconco™ Protector XVS™ Ventilation Stations: PVC Duct
Accessories for Labconco Protector XVS Ventilation Stations
Brand:  Labconco™ 4708600
Product Code. 15489434
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use with Labconco Protector XVS Ventilation Stations
Ductwork
  • Standard 6 in. dia. (15.2cm) Type I PVC duct can be cut without special tools; secured with solvent cement (Female duct coupling required to join two sections. See Cat. No. 16-304-12)
  • 90° elbow (Cat. No. 16-304-11) has belled end connections to receive PVC duct directly
  • Flexible duct connection reduces vibration between blower and PVC ductwork; supplied with two installation clamps
  • Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
  • PVC zero-pressure weathercap (Cat. No. 16-304-14) connects directly to the duct for vertical discharge of effluent air and dispersion away from the building
  • Cap keeps out rain and snow but adds little static pressure to the exhaust system
Specifications
Specifications
Protector XVS Ventilation Station
11.3 Kg
PVC
25 lb.
PVC Duct
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Protector XVS™ Ventilation Stations: PVC Duct > PVC Duct