Learn More
Labconco™ Purifier™ Logic+ Class II B2 Biosafety Cabinets, 4 ft. Width, 8 in. Opening, International Model
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
Brand: Labconco™ 303481150
Additional Details : Weight : 10.00000kg
Description
MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.
- ECM blower technology combined with CAP (Constant Airflow Profile™) maintains precise volume of air as filters load
- Interior-mounted LCD displays HEPA filter life remaining and cabinet status
- Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
- Cabinets are designed to be hard ducted to the outside
- Flush-mounted stainless steel electrical receptacle covers with dampening hinges
- Patented curved air grille with built-in armrest
- Fully accessible controls and eye-level mounted color display with MyLogic operating system
Some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. (Refer to nominal width, sash opening, electrical configuration and product specifications for details.)
Specifications
ADA, CE, NSF 49 | |
UK (230 V) | |
20.3 cm | |
Powder Coated Steel Exterior and Stainless Steel Interior | |
Fluorescent, UV | |
54.3 in. | |
27 in. | |
79.2 cm | |
331.1 Kg | |
6 A |
Stainless Steel | |
100.1 to 106.1 in. | |
LCD | |
Base Stand, UV Light, Service Fixtures, Pass-through | |
254.2 to 269.2 cm | |
137.9 cm | |
31.2 in. | |
730 lb. | |
230 V |