missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Purifier™ Logic+ Class II B2 Biosafety Cabinets, 4 ft. Width, 8 in. Opening, International Model
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
£11321.20
Specifications
Chamber Material | Stainless Steel |
---|---|
Sash Opening (Metric) | 20.3 cm |
Display | LCD |
Housing Material | Powder Coated Steel Exterior and Stainless Steel Interior |
Width (English) | 54.3 in. |
Product Code | Brand | Height (Metric) | Includes | Lighting | Plug Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Includes | Lighting | Plug Type | Price | Quantity & Availability | ||
18044122
|
Labconco™
303481150 |
254.2 to 269.2 cm | Base Stand, UV Light, Service Fixtures, Pass-through | Fluorescent, UV | UK (230 V) |
£11321.20
Each |
Estimated Shipment: 18-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.
- ECM blower technology combined with CAP (Constant Airflow Profile™) maintains precise volume of air as filters load
- Interior-mounted LCD displays HEPA filter life remaining and cabinet status
- Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
- Cabinets are designed to be hard ducted to the outside
- Flush-mounted stainless steel electrical receptacle covers with dampening hinges
- Patented curved air grille with built-in armrest
- Fully accessible controls and eye-level mounted color display with MyLogic operating system
Some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. (Refer to nominal width, sash opening, electrical configuration and product specifications for details.)
Specifications
Stainless Steel | |
LCD | |
54.3 in. | |
27 in. | |
79.2 cm |
20.3 cm | |
Powder Coated Steel Exterior and Stainless Steel Interior | |
137.9 cm | |
31.2 in. |