missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Lab Washer PG 8504 AE AD

Optimally designed laboratory washer with perfectly aligned wash pressure and ideally designed baskets, filters, and spray arms. Miele™ PG 8504 AE AD Laboratory Glassware Washer features a variety of specific wash programs to meet multiple laboratory needs.

Brand:  Miele Professional™ 62850421

5363.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15427694

  • £10730.00 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Variable-speed heater pump
  • Optimum cleaning performance
  • Optimum wash pressure
  • Low consumption values
  • Short batch times
  • Crevice-free wash cabinet
  • De-min. water connection
Specifications

Specifications

Freestanding or built-under
500 L/min.
De-min. water connection, electrical door lock, acoustic end of program signal, integrated water softener
600 mm
835 mm
3 AC 208V 60HZ
Stainless Steel
IP safety class according to EN 60529: IP 21, EN 61010-1, EN 61010-2-040, RoHS Directive 2011/65/EU, c CSA us
70°C
474/520 x 530 x 520mm
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Miele Professional™ Lab Washer PG 8504 AE AD > Includes: De-min. water connection, electrical door lock, acoustic end of program signal, integrated water softener

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.