Learn More
Cytiva Laboratory Manifold 5/16 in. Hose Barb End Cap
Replacement tubing outlet accessory for microbiology manifold.
Brand:  Cytiva 4887
Product Code. 17194431
  • £289.00 / Pack of 2
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laboratory Manifold
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Laboratory Manifold 5/16 in. Hose Barb End Cap > For use with Laboratory Manifold