missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Nickel Crucible Lids

Obtain optimum success with nickel crucible lids during the heating process preventing the chance of hot material spattering out of an open crucible. In fusions where zirconium or other metals cannot be used, the Clifton™ Nickel Crucible Lids are the ideal alternative.

Brand:  Nickel Electro™ 47

 View more versions of this product

Product Code. 11479579

  • £12.75 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Provides a cover to crucibles
  • Prevents chance of hot material spattering out of open crucible during heating process
  • Lid can be easily removed with either stainless steel or nickel laboratory tongs 
  • Forms a tightly adhering oxide file between 400°C and 600°C in oxidizing atmospheres
  • Nickel effective for use in fusions where zirconium or other metals cannot be used
Specifications

Specifications

Nickel
Each
Laboratory Crucible Lid
350°C
35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Electro™ Clifton™ Nickel Crucible Lids > Diameter: 35mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.