missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 14cm Diamter

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 14cm Diameter offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

POA

Specifications

Dimensions (L x W) 14 x 14 cm
Type Lab Safety Mat
Quantity 50/Pk.
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15737069
View Documents Promotion Details
Dycem™
NS2141
Blue
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15747069
View Documents Promotion Details
Dycem™
LS02142
Red
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15757069
View Documents Promotion Details
Dycem™
LS02144
Silver
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15767069
View Documents Promotion Details
Dycem™
LS02146
Black
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15777069
View Documents Promotion Details
Dycem™
LS02148
Yellow
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Anti-slip properties
  • 14cm diameter
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors
Specifications

Specifications

14 x 14 cm
Lab Safety Mat
50/Pk.
Videos
SDS
Documents

Documents

Product Certifications