missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 25 x 18cm

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 25 x 18cm offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

Brand:  Dycem™ NS02SM4

Additional Details : Weight : 0.21000kg

 View more versions of this product

Product Code. 15737089

  • POA / Pack of 50
Estimated Shipment: 10-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Anti-slip properties
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors
Specifications

Specifications

25 x 18 cm
Silver
Lab Safety Mat
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers