missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dycem™ Laboratory Safety Mats 25 x 18cm
Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 25 x 18cm offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.
Brand: Dycem™ NS02SM8
This item is not returnable.
View return policy
Description
- Anti-slip properties
- Ideal for use where spillage or damage can occur
- Reduce movement and provide bench protection
- Protect technicians and equipment
- Available in a range of colors
Specifications
25 x 18 cm | |
50/Pk. | |
Yellow | |
Lab Safety Mat |