missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dycem™ Laboratory Safety Self-Adhesive Discs
Effectively increase the gripping properties of all surfaces. Dycem™ Laboratory Safety Self-Adhesive Discs are ideal for securing items to create a more permanent non-slip surface.
Brand: Dycem™ LS05196
Additional Details : Weight : 0.20000kg
Description
- Can be used with furniture
- Improves grip and safety
- Small and discrete
Specifications
19mm Diameter x 3m | |
Self-adhesive Nonslip Disc | |
50/Pk. |
Black | |
19 mm | |
3 mm |