missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Rabbit Polyclonal Antibody, Epredia™

One marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available is the Epredia™ Lambda Light Chain, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-334-A0

 View more versions of this product

Product Code. 12683227

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Mouse and Rat. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Lambda light chains purified from human Bence Jones proteins

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Lambda Light Chain
Polyclonal
Rabbit
100 μL
Hematopoietic
Human, Non-human Primate, Rat
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Lambda light chains purified from human Bence Jones proteins
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.