Learn More
Lambda Light Chain Rabbit Polyclonal Antibody, Epredia™
One marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available is the Epredia™ Lambda Light Chain, Rabbit Polyclonal Antibody.
£122.40 - £383.00
Specifications
Antigen | Lambda Light Chain |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12683227
|
Epredia
RB-334-A0 |
Concentrated | 100 μL | N/A | |||||
12693227
|
Epredia
RB-334-A1 |
Concentrated | 500 μL |
£252.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12673227
|
Epredia
RB-334-A |
Concentrated | 1 mL |
£383.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12623237
|
Epredia
RB-334-R7 |
Ready-to-Use | 7 mL | N/A | |||||
12633237
|
Epredia
RB-334-RQ |
Ready-to-Use | 12 mL | N/A | |||||
Description
Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Baboon, Mouse and Rat. Does not react with dog. Others not known.
Epitope: Not determined
Immunogen: Lambda light chains purified from human Bence Jones proteins
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Lambda Light Chain | |
Polyclonal | |
Rabbit | |
Hematopoietic | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Lambda light chains purified from human Bence Jones proteins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.