missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminated Polyester Warning Signs
£7.65 - £89.57
Specifications
Material | Polyester |
---|---|
Adhesive Type | Permanent Acrylic |
Description | Laminated Polyester |
For Use With (Application) | Pipe markers and safety signs |
Thickness | 0.024 mm |
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
10403194
|
Brady™
250264 |
Warning of Hazardous Substances | 100 mm |
£7.65
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
15138743
|
Brady™
227360 |
Radioactive substances 7C III | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Polyester | |
Laminated Polyester | |
0.024 mm | |
Warning Sign |
Permanent Acrylic | |
Pipe markers and safety signs | |
Triangular | |
Black on Yellow |