missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Laminated Silicone Rubber Septa
Brand: Hamilton™ 76015
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
3 | |
Laminated Silicone Rubber | |
Septum |
14 mm | |
200°C | |
12/Pk. |