missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Laminated Silicone Rubber Septa
Brand: Hamilton™ 76013
Code : A0
Additional Details : Weight : 0.01000kg
Specifications
3 | |
Laminated Silicone Rubber | |
Septum |
12 mm | |
200°C | |
12/Pk. |