Learn More
Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts. Due to its dielectric properties it is also posses properties for low-loss microwave and dielectric resonance applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 25 g |
Solubility Information | Soluble in phosphorous acid (at >150°C). Insoluble in mineral acid ( at 25°C) and water. |
Formula Weight | 213.88 |
Percent Purity | 99.9% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Lanthanum aluminum oxide |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W13K033 | Certificate of Analysis | 25/09/2024 | 039250.06,039250.14 |
P20K015 | Certificate of Analysis | 02/04/2024 | 039250.06,039250.14 |
R08J008 | Certificate of Analysis | 20/02/2024 | 039250.06,039250.14 |
W23F060 | Certificate of Analysis | 18/11/2021 | 039250.06,039250.14 |
H02Q34 | Certificate of Analysis | 18/11/2021 | 039250.06,039250.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.