missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum chloride, 99.9%, Thermo Scientific Chemicals Product Code.: 10404545

Lanthanum chloride, 99.9%, Thermo Scientific Chemicals

Product Code. 10404545
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10404545

Brand: Thermo Scientific Acros 315230500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Boiling Point 1000.0°C
Melting Point 860.0°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9%
Linear Formula LaCl3
Merck Index 15,5414
Solubility Information Solubility in water: soluble
Formula Weight 245.26
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Lanthanum chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462526 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number315230500,315230100,315232500
Lot NumberA0453542 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number315230500,315230100,315232500
Lot NumberA0451933 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number315230500,315230100,315232500
Lot NumberA0448926 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number315230500,315230100,315232500
Lot NumberA0402827 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number315230500,315230100,315232500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum chloride, 99.9%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.