missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals

£44.22 - £703.50

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11483047
View Documents
Thermo Scientific Alfa Aesar
A15575.22
100 g
£44.22
100g
Please to purchase this item. Need a web account? Register with us today!
11493047
View Documents
Thermo Scientific Alfa Aesar
A15575.36
500 g
£175.88
500g
Please to purchase this item. Need a web account? Register with us today!
11403057
View Documents
Thermo Scientific Alfa Aesar
A15575.0E
2500 g
£703.50
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

Solubility
Soluble in ethanol and water.

Notes
Incompatible with oxidizing agents, strong acids, strong reducing agents and active metals.
Specifications

Chemical Identifiers

10025-84-0
371.36
FDFPDGIMPRFRJP-UHFFFAOYSA-K
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
Cl3H14LaO7
MFCD00149756
lanthanum(3+) heptahydrate trichloride

Specifications

91°C (decomposition)
Cl3H14LaO7
14,5363
FDFPDGIMPRFRJP-UHFFFAOYSA-K
lanthanum(3+) heptahydrate trichloride
371.37 (245.27 Anhydrous)
Odorless
10025-84-0
MFCD00149756
Soluble in ethanol and water.
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
371.36
99%
Lanthanum(III) chloride heptahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H317-H318

missing translation for 'einecsNumber' : 233-237-5

missing translation for 'rtecsNumber' : OE4470000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lanthanum(III) chloride heptahydrate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Lanthanum(III) chloride heptahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only