Learn More
Lanthanum(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Lanthanum(III) fluoride is used in phosphor lamp coating, carbon arc electrodes and in lasers. It is also used in fluoride selective electrodes. Further, it is used as a non-ferrous metal additive and used in metal alloys.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1493°C |
Color | White |
Physical Form | Crystals |
Quantity | 50 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Soluble in strong mineral acids. Insoluble in water. |
Formula Weight | 195.9 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M04K004 | Certificate of Analysis | 18/01/2024 | 011273.18,011273.30 |
X16J055 | Certificate of Analysis | 13/11/2023 | 011273.18,011273.30 |
S20J046 | Certificate of Analysis | 27/06/2023 | 011273.18,011273,011273.30 |
M08H023 | Certificate of Analysis | 12/01/2023 | 011273.18,011273.30 |
T06H021 | Certificate of Analysis | 12/11/2021 | 011273.18,011273.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.