Learn More
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
£102.00 - £343.00
Chemical Identifiers
CAS | 10277-43-7 |
---|---|
Molecular Formula | H12LaN3O15 |
Molecular Weight (g/mol) | 433.01 |
MDL Number | MFCD00149751 |
InChI Key | GJKFIJKSBFYMQK-UHFFFAOYSA-N |
Synonym | lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate |
PubChem CID | 16211478 |
IUPAC Name | lanthanum(3+) hexahydrate trinitrate |
SMILES | O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10607192
|
Thermo Scientific Acros
218711000 |
100 g | Glass bottle |
£102.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10571081
|
Thermo Scientific Acros
218715000 |
500 g | Glass bottle |
£343.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10277-43-7 | |
433.01 | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
16211478 | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12LaN3O15 | |
MFCD00149751 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
lanthanum(3+) hexahydrate trinitrate |
Specifications
126.0°C | |
10277-43-7 | |
Powder | |
H12LaN3O15 | |
MFCD00149751 | |
15, 5414 | |
Solubility in water: 1580g/L (25°C). Other solubilities: soluble in acids, soluble in alcohol | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
433.01 | |
433.01 | |
Lanthanum(III) nitrate hexahydrate, 98+% |
40.0°C | |
White to Yellow | |
98% min. (Complexometry) | |
La(NO3)3·6H2O | |
11,283 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
lanthanum(3+) hexahydrate trinitrate | |
16211478 | |
≥98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : OE5250000
RUO – Research Use Only