missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11376338

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11376338
5 g
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11376338

Brand: Thermo Scientific Alfa Aesar 011267.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronateShow MoreShow Less
PubChem CID 16211478
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 40°C
Color White
Physical Form Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water,alcohol and acids.
IUPAC Name lanthanum(3+) hexahydrate trinitrate
Formula Weight 433.01 (324.92 Anhydrous)
Odor Odorless
Appearance White crystalline
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ12K006 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number011267.06,011267.18,011267.PG,011267.30,011267.A1
Lot NumberR22K002 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011267.06,011267.18,011267.PG,011267.30,011267.A1
Lot NumberR17K017 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011267.06,011267.18,011267.PG,011267.30,011267.A1
Lot NumberN19K010 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number011267.06,011267.18,011267.PG,011267.30,011267.A1
Lot NumberY01J033 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number011267.06,011267.18,011267.PG,011267.30,011267.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.