Learn More
Lanthanum(III) nitrate hexahydrate, REacton™, 99.999% (REO), Thermo Scientific Chemicals
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 40°C |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.999% (REO) |
Solubility Information | Soluble in water,alcohol and acids. |
IUPAC Name | lanthanum(3+) hexahydrate trinitrate |
Formula Weight | 433.01 (324.92 Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X31K010 | Certificate of Analysis | 14/11/2024 | 044346.22,044346.14,044346.36 |
X13J038 | Certificate of Analysis | 24/10/2023 | 044346.22,044346.14,044346.36 |
X13J039 | Certificate of Analysis | 24/10/2023 | 044346.22,044346.14,044346.36 |
Z14I024 | Certificate of Analysis | 13/01/2023 | 044346.22,044346.14,044346.36 |
Z30I019 | Certificate of Analysis | 10/01/2023 | 044346.22,044346.14,044346.36 |
Safety and Handling


- Lanthanum(III) nitrate hexahydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Lanthanum(III) nitrate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.