missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11386328

Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11386328
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11386328

Brand: Thermo Scientific Alfa Aesar 011264.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071

Specifications

Boiling Point 4200°C
Melting Point 2307°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water. Soluble in dilute acid.
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Density 6.51
Chemical Name or Material Lanthanum(III) oxide, Rare Earth Oxides Content
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP21L030 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog Number011264.18,011264.30,011264.A1
Lot NumberM12K041 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number011264.18,011264.30,011264.A1
Lot NumberN14J053 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011264.18,011264.30,011264.A1
Lot NumberZ23G039 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011264.18,011264.30,011264.A1
Lot NumberN18I007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011264.18,011264.30,011264.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.