missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Eu-PY20 Antibody Kit
LanthaScreen™ Eu-PY20 Antibody Kit
Brand: Thermo Scientific™ PV5692
Code : 50
Additional Details : Weight : 0.10000kg
Description
A general reagent to monitor tyrosine kinase activity, the PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate. For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications.Specifications
Tyrosine kinases | |
0.53mg/mL | |
150kDa | |
RUO, RUO | |
Primary | |
Europium-labeled anti-PY20 antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
Fluorescent | |
Dry Ice | |
TR-FRET | |
LanthaScreen™ |
Kinase Assay, Kinase Assay | |
Europium | |
25 μg | |
Pharma and Biopharma | |
1 Kit(s) | |
Biochemical europium phospho-specific activity | |
340⁄615 | |
ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA V561D, SYK, TYRO3, ZAP70, FGFR1, FGFR2, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, TEK, FLT1, MUSK, SRCN1, ABL1 E255K, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, EPHA7, EPHB1, FRK, ITK, FLT3 D835Y, SRC, BTK, FES, ERBB2, NTRK2 | |
EGFR |
For Research Use Only. Not for use in diagnostic procedures.