missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Tb-DUB Substrate
LanthaScreen™ Tb-DUB Substrate
£337.00 - £2750.00
Specifications
For Use With (Application) | Pharma and biopharma, target and lead identification and validation, drug discovery and development, proteasome biology, biochemical proteasome assays, epigenetics assays for drug discovery |
---|---|
Substrate Type | Deconjugating enzyme substrate |
Molecular Weight (g/mol) | 42.8kDa |
Product Type | LanthaScreen™ Tb-DUB Substrate |
Storage Requirements | –80°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11515046
|
Invitrogen
PV4444 |
100μg |
£2750.00
100µg |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
11505046
|
Invitrogen
PV4443 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Deubiquitinating enzymes (DUBs) play housekeeping functions by maintaining pools of active ubiquitin, and regulatory function by rescuing specific target proteins from degradation by the proteasome. The LanthaScreen™ Tb-DUB Substrate is a general substrate for assaying DUB activity in a high throughput assay format.Specifications
Pharma and biopharma, target and lead identification and validation, drug discovery and development, proteasome biology, biochemical proteasome assays, epigenetics assays for drug discovery | |
42.8kDa | |
–80°C |
Deconjugating enzyme substrate | |
LanthaScreen™ Tb-DUB Substrate |
For Research Use Only. Not for use in diagnostic procedures.