Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Ab
LanthaScreen™ Tb-labeled Anti-Mouse Ab
Brand:  Invitrogen™ PV3768
Code : 50
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 10003574
  • £10240.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • A general reagent for use in assay development
  • This terbium labeled antibody will bind mouse antibodies
  • It is ideally suited for use during assay development when evaluating multiple primary mouse antibodies
  • Also ideal for use when it is impossible to directly terbium label a primary antibody
Specifications
Specifications
Lanthanide Chelates, Terbium (Tb)
Mouse
End Point
1 mg
Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C.
Dry Ice
340/490, 520
LanthaScreen™