Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Ab
LanthaScreen™ Tb-labeled Anti-Mouse Ab
Brand:  Invitrogen™ PV3766
 View more versions of this product
Product Code. 10309204
  • £752.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
A general reagent for use in assay development, this terbium labeled antibody will bind mouse antibodies. This product is ideally suited for use during assay development when evaluating multiple primary mouse antibodies or when you are unable to directly terbium label your primary antibody.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C.
25 μg
End Point
Lanthanide Chelates, Terbium (Tb)
340/490, 520
Mouse
Dry Ice