missing translation for 'onlineSavingsMsg'
Learn More

MakerBot™ Large Translucent PLA Filament

MakerBot™ Translucent PLA Filament is composed of the purest grade resin and designed for use with the MakerBot™ 2 Desktop 3D printer and all fifth generation MakerBot™ Replicator 3D printers.

POA

Specifications

For Use With (Equipment) Replicator 3D Printer, Replicator 2 3D Printer, Replicator+ 3D Printer, Replicator Z18 3D Printer
Product Type PLA Filament
Material PLA
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16321496
Encompass_Preferred
View Documents
MakerBot™
MKS-000016
Translucent Red
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16331496
Encompass_Preferred
View Documents
MakerBot™
MKS-000017
Translucent Orange
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16341496
Encompass_Preferred
View Documents
MakerBot™
MKS-000019
Translucent Green
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16351496
Encompass_Preferred
View Documents
MakerBot™
MKS-000020
Translucent Blue
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16361496
Encompass_Preferred
View Documents
MakerBot™
MKS-000021
Translucent Purple
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Composed of the purest grade resin on the market
  • Comprehensively quality tested
  • Reduces downtime
  • Creates high quality prints
  • Filament Diameter: 1.75 mm
  • Applications: CAD, Conceptual Modeling, Prototyping, Education, AEC
  • Packaging Dimensions (L x W x H): 250 x 250 x 40 mm
Specifications

Specifications

Replicator 3D Printer, Replicator 2 3D Printer, Replicator+ 3D Printer, Replicator Z18 3D Printer
PLA Filament
PLA
Videos
SDS
Documents

Documents

Product Certifications