Learn More
Motic™ LD Plan Achromatic Objectives
Plan Achromatic objectives for use with TEC POL Microscopes.
Brand:  Motic™ 1101000203621
 View more versions of this product
Product Code. 17188598
  • £241.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Motic™ Panthera TEC POL Microscopes
  • LD Plan Achromatic objectives
Specifications
Specifications
Microscope Objective
Plan Achromatic
0.25
Motic™ TEC POL Microscopes
10X
17.5 mm