missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE
Designed to give an accurate fine controlled stream of liquid. Kautex™ 500mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.
Brand: Kautex™ 2000770021
Code : NEW
Additional Details : Weight : 0.33840kg
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
500 mL | |
39.2 mm | |
10 | |
Green | |
Round | |
Wash Bottle | |
77 mm |
Natural | |
LDPE | |
100 Pk. | |
Wide | |
154 mm | |
Ethanol | |
Screw Cap with Delivery Nozzle |