missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals Product Code.: 10035683

Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Product Code. 10035683
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10035683

Brand: Thermo Scientific Acros 317230050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrateShow MoreShow Less
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula (CH3CO2)2Pb·3H2O
Fieser 01,532; 02,233; 04,276
Merck Index 15, 5452
Solubility Information Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
IUPAC Name λ²-lead(2+) diacetate trihydrate
Formula Weight 379.33
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lead(II) acetate trihydrate, 99.999%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457771 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number317230050,317230250
Lot NumberA0452385 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog Number317230050,317230250
Lot NumberA0397659 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number317230050,317230250
Lot NumberA0419875 Certificate TypeCertificate of Analysis Date08/12/2022 Catalog Number317230050,317230250
Lot NumberA0436133 Certificate TypeCertificate of Analysis Date10/11/2021 Catalog Number317230050,317230250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) acetate trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.