Learn More
Lead(II) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 916°C |
Melting Point | 373°C |
Color | White |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.999% (Metals basis) |
Solubility Information | Slightly soluble in water. Insoluble in alcohol. Soluble in ammonia and alkali. |
IUPAC Name | dibromolead |
Formula Weight | 367.01 |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P11L025 | Certificate of Analysis | 17/03/2025 | 035703.14,035703.03,035703.06 |
10253540 | Certificate of Analysis | 08/01/2025 | 035703.03,035703.14,035703.06 |
10253539 | Certificate of Analysis | 08/01/2025 | 035703.03,035703.14,035703.06 |
17318 | Certificate of Analysis | 05/09/2024 | 035703.03,035703.14,035703.06 |
X26H058 | Certificate of Analysis | 12/04/2022 | 035703.03,035703.14,035703.06 |
Safety and Handling



- Lead(II) bromide
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.