missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

£87.20 - £679.00

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
SMILES [Br-].[Br-].[Pb++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035703.03
1 g
£87.20
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11388537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035703.06
5 g
£138.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11398537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035703.14
25 g
£679.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
[Br-].[Br-].[Pb++]

Specifications

916°C
10031-22-8
Powder
Br2Pb
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
Odorless
Lead(II) bromide
373°C
White
99.999% (Metals basis)
MFCD00011156
Slightly soluble in water. Insoluble in alcohol. Soluble in ammonia and alkali.
[Br-].[Br-].[Pb++]
367.00
367.01
6.66
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c

H302+H332-H360Df-H373

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 233-084-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only