Learn More
Lead(II) carbonate, ACS reagent, Thermo Scientific Chemicals
£37.29 - £122.61
Chemical Identifiers
CAS | 598-63-0 |
---|---|
Molecular Formula | CO3Pb |
Molecular Weight (g/mol) | 267.20 |
InChI Key | MFEVGQHCNVXMER-UHFFFAOYSA-L |
Synonym | lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 |
PubChem CID | 11727 |
SMILES | [Pb++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12654957
|
Thermo Scientific Acros
212511000 |
100 g |
£37.29
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12644957
|
Thermo Scientific Acros
212515000 |
500 g |
£122.61
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-63-0 | |
267.20 | |
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 | |
[Pb++].[O-]C([O-])=O |
CO3Pb | |
MFEVGQHCNVXMER-UHFFFAOYSA-L | |
11727 |
Specifications
400.0°C | |
CO3Pb | |
10,1006 | |
Solubility in water: insoluble | |
[Pb++].[O-]C([O-])=O | |
267.20 | |
267.2 | |
Plastic bottle |
598-63-0 | |
PbCO3 | |
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 | |
MFEVGQHCNVXMER-UHFFFAOYSA-L | |
λ²-lead(2+) carbonate | |
11727 | |
ACS Reagent | |
Lead(II) carbonate |
Safety and Handling
GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-943-4
- Lead(II) carbonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only