Learn More
Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals
Analytical Reagent, Preparation of Lead Salts, As Solder and Flux
£23.31 - £139.70
Chemical Identifiers
CAS | 7758-95-4 |
---|---|
Molecular Formula | Cl2Pb |
Molecular Weight (g/mol) | 278.10 |
MDL Number | MFCD00011157 |
InChI Key | HWSZZLVAJGOAAY-UHFFFAOYSA-L |
Synonym | lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dryShow More |
PubChem CID | 24459 |
IUPAC Name | dichlorolead |
SMILES | [Cl-].[Cl-].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371719
|
Thermo Scientific Alfa Aesar
012349.18 |
50 g |
£23.31
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381719
|
Thermo Scientific Alfa Aesar
012349.36 |
500 g |
£56.23
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361719
|
Thermo Scientific Alfa Aesar
012349.A3 |
2 kg |
£139.70
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed in the manufacture of infrared transmitting glass, and ornamental glass called arena glass. It also finds application as analytical reagent, in preparation of lead salts, as solder and flux, for provision of lead titanate, and used in the purification process of ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-95-4 | |
278.10 | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
24459 | |
[Cl-].[Cl-].[Pb++] |
Cl2Pb | |
MFCD00011157 | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dryShow More | |
dichlorolead |
Specifications
951°C | |
7758-95-4 | |
Crystalline Powder | |
Cl2Pb | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
dichlorolead | |
24459 | |
Reagent | |
5.85 g/cm3 at 20°C |
501°C | |
White | |
99% | |
MFCD00011157 | |
Soluble in hot water,alkali hydroxides and NH4Cl solution. Insoluble in cold water and alcohol. | |
[Cl-].[Cl-].[Pb++] | |
278.10 | |
278.1 | |
Odorless | |
Lead(II) chloride |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-845-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.