missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11328547

Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11328547
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11328547

Brand: Thermo Scientific Alfa Aesar 035704.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells.

Lead(II) chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dryShow MoreShow Less
PubChem CID 24459
IUPAC Name dichlorolead
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Boiling Point 951°C
Melting Point 501°C
Physical Form Beads
Quantity 5 g
UN Number UN2291
Solubility Information Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol.
Formula Weight 278.1
Percent Purity 99.999%
Packaging Ampouled under argon
Specific Gravity 5.85
Density 5.85
Chemical Name or Material Lead(II) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX11J034 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number035704.22,035704.06,035704.14
Lot NumberS22J007 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number035704.22,035704,035704.06,035704.14
Lot NumberH16S005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035704.22,035704.06,035704.14
Lot NumberG02N35 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035704.22,035704.06,035704.14
Lot NumberT24B010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035704.22,035704.06,035704.14
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) chloride, ultra dry
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.