missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) formate, 90+%, Thermo Scientific™

£20.70 - £229.00

Chemical Identifiers

CAS 811-54-1
Molecular Formula C2H2O4Pb
Molecular Weight (g/mol) 297.234
MDL Number MFCD00050834
InChI Key ZGVSCHRFRACEIO-UHFFFAOYSA-L
Synonym lead diformate, lead formate, formic acid, lead 2+ salt, unii-2kjp65ymf4, lead ii formate, lead 2+ formate, 2kjp65ymf4, lead formate pb o2ch 2, lead formate pb hco2 2, formic acid, lead 2+ salt 2:1
PubChem CID 61216
IUPAC Name lead(2+);diformate
SMILES C(=O)[O-].C(=O)[O-].[Pb+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14357.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11456927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14357.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11436927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14357.0B
1000 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

811-54-1
297.234
ZGVSCHRFRACEIO-UHFFFAOYSA-L
61216
C(=O)[O-].C(=O)[O-].[Pb+2]
C2H2O4Pb
MFCD00050834
lead diformate, lead formate, formic acid, lead 2+ salt, unii-2kjp65ymf4, lead ii formate, lead 2+ formate, 2kjp65ymf4, lead formate pb o2ch 2, lead formate pb hco2 2, formic acid, lead 2+ salt 2:1
lead(2+);diformate

Specifications

190°C (decomposition)
C2H2O4Pb
UN2291
lead diformate, lead formate, formic acid, lead 2+ salt, unii-2kjp65ymf4, lead ii formate, lead 2+ formate, 2kjp65ymf4, lead formate pb o2ch 2, lead formate pb hco2 2, formic acid, lead 2+ salt 2:1
C(=O)[O-].C(=O)[O-].[Pb+2]
297.234
297.23
Lead(II) formate
811-54-1
MFCD00050834
14,5409
ZGVSCHRFRACEIO-UHFFFAOYSA-L
lead(2+);diformate
61216
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications