missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals Product Code.: 10551364

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals

Product Code. 10551364
2.5 kg, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10551364

Brand: Thermo Scientific Acros 315850025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Boiling Point 1470.0°C
Melting Point 886.0°C
Color Yellow
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99.9+%
Linear Formula PbO
Fieser 01,536; 02,233
Merck Index 15, 5468
Solubility Information Solubility in water: 70.2mg/l (20°C)
IUPAC Name plumbanone
Formula Weight 223.19
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lead(II) oxide, <10 micron, 99.9+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461472 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number315850025,315851000,315855000
Lot NumberA0452327 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number315850025,315851000,315855000
Lot NumberA0392873 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number315850025,315851000,315855000
Lot NumberA0387512 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number315850025,315851000,315855000
Lot NumberA0442074 Certificate TypeCertificate of Analysis Date14/05/2022 Catalog Number315850025,315851000,315855000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.