missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) perchlorate trihydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 100.0°C |
Color | White to Beige |
pH | 3 to 5 (5% soln.) |
Physical Form | Adhering Crystals |
Quantity | 25 g |
Assay Percent Range | 98.5% min. (Complexometry) |
Linear Formula | Pb(ClO4)2·3H2O |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol |
IUPAC Name | lead(2+);diperchlorate;trihydrate |
Formula Weight | 460.14 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458773 | Certificate of Analysis | 31/01/2025 | 254592500,254590250,254590010 |
A0450593 | Certificate of Analysis | 27/02/2024 | 254592500,254590250,254590010 |
A0458773 | Certificate of Analysis | 08/01/2024 | 254592500,254590250,254590010 |
A0400836 | Certificate of Analysis | 13/12/2022 | 254592500,254590250,254590010 |
A0445316 | Certificate of Analysis | 02/09/2022 | 254592500,254590250,254590010 |
Safety and Handling




Product Identifier
- Lead(II) perchlorate trihydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lead(II) perchlorate trihydrate, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction