missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11490051

Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11490051
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11490051

Brand: Thermo Scientific Alfa Aesar 044784.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Pigments, Galvanic Batteries, Lithography

Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.256
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesiteShow MoreShow Less
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170°C
Color White
Physical Form Lump/Powder
Quantity 100 g
Solubility Information Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol
Formula Weight 303.25
Grade Reagent
Odor Odorless
Appearance White powder/lump
Density 6.2 g/cm3 at 20°C
Chemical Name or Material Lead(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU30K010 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number044784.36,044784.22
Lot NumberM12K003 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number044784.36,044784.22
Lot NumberR17F052 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number044784.36,044784.22
Lot NumberU09J046 Certificate TypeCertificate of Analysis Date21/08/2023 Catalog Number044784.36,044784.22
Lot NumberR23I003 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number044784.36,044784.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.