missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) sulfide, 90%, pure, Thermo Scientific Chemicals Product Code.: 15266048

Lead(II) sulfide, 90%, pure, Thermo Scientific Chemicals

Product Code. 15266048
5 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 15266048

Brand: Thermo Scientific Acros 453970050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfideShow MoreShow Less
PubChem CID 14819
IUPAC Name λ²-lead(2+) sulfanediide
SMILES [S--].[Pb++]

Specifications

Boiling Point 1281.1°C
Melting Point 1113.9°C
Quantity 5 kg
Assay Percent Range 78% min. (Pb)
Merck Index 15,5476
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in nitric acid
Formula Weight 239.26
Percent Purity 90%
Grade Pure
Chemical Name or Material Lead(II) sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466599 Certificate TypeCertificate of Analysis Date04/10/2024 Catalog Number453970010,453970050
Lot NumberA0432324 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog Number453970010,453970050
Lot NumberA0447419 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog Number453970010,453970050
Lot NumberA0441343 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog Number453970010,453970050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) sulfide, 90%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.