missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfide, 90%, pure, Thermo Scientific Chemicals

£93.67 - £434.16

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name λ²-lead(2+) sulfanediide
SMILES [S--].[Pb++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15256048
View Documents
Thermo Scientific Acros
453970010
1 kg
£93.67
1kg
Please to purchase this item. Need a web account? Register with us today!
15266048
View Documents
Thermo Scientific Acros
453970050
5 kg
£434.16
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-87-0
239.30
FILXVLQXZYXBSJ-UHFFFAOYSA-N
14819
[S--].[Pb++]
PbS
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
λ²-lead(2+) sulfanediide

Specifications

1281.1°C
1314-87-0
PbS
15,5476
Solubility in water: insoluble. Other solubilities: soluble in nitric acid
[S--].[Pb++]
239.30
239.26
Pure
1113.9°C
78% min. (Pb)
MFCD00016280
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
FILXVLQXZYXBSJ-UHFFFAOYSA-N
λ²-lead(2+) sulfanediide
14819
90%
Lead(II) sulfide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquati

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 215-246-6

Product Identifier
  • Lead(II) sulfide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only