Learn More
Lead(II) sulfide, Pb 82% min., Thermo Scientific Chemicals
Description
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.
Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 1114°C |
Quantity | 500 g |
UN Number | UN3077 |
Merck Index | 14,5421 |
Solubility Information | Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide. |
IUPAC Name | λ²-lead(2+) sulfanediide |
Formula Weight | 239.25 |
Concentration or Composition (by Analyte or Components) | Pb 82% min. |
Odor | Odorless |
Refractive Index | 3.921 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0447419 | Certificate of Analysis | 30/08/2024 | A10268.0E,A10268.36 |
10219870 | Certificate of Analysis | 25/12/2023 | A10268.0E,A10268.36 |
B6761A | Certificate of Analysis | 22/11/2021 | A10268.0E,A10268.36 |
10182147 | Certificate of Analysis | 22/11/2021 | A10268.0E,A10268.36 |
10176230 | Certificate of Analysis | 22/11/2021 | A10268.0E,A10268.36 |
Safety and Handling



- Lead(II) sulphide
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Lead(II) sulphide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.