Learn More
Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals
Description
It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Brown to Black |
Physical Form | Powder |
Quantity | 2 kg |
Assay Percent Range | 97.0% min. |
IUPAC Name | dioxolead |
Formula Weight | 239.19 |
Grade | ACS |
Odor | Odorless |
Chemical Name or Material | Lead(IV) oxide |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P24L020 | Certificate of Analysis | 26/03/2025 | 012483.36,012483.A3,012483.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.