Learn More
Lead wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Appearance | Gray wire |
Density | 11.34g/cm3 at 20°C |
Assay Percent Range | 99.998% (Metals basis) |
Quantity | 1 m |
Odor | Odorless |
Form | Wire |
Chemical Name or Material | Lead |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T25I016 | Certificate of Analysis | 02/08/2022 | 011901.G1,011901.G5 |
Y20G041 | Certificate of Analysis | 08/12/2021 | 011901.G1,011901.G5 |
Q27D026 | Certificate of Analysis | 29/11/2021 | 011901.G1,011901.G5 |
U10H012 | Certificate of Analysis | 29/11/2021 | 011901.G1,011901.G5 |
P19E063 | Certificate of Analysis | 24/11/2021 | 011901.G1,011901.G5 |
Safety and Handling



- Lead wire
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.