missing translation for 'onlineSavingsMsg'
Learn More
Lead wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11391998

Lead wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11391998
5 m
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy

Product Code. 11391998

Brand: Thermo Scientific Alfa Aesar 011901.G5

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.8.9g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Appearance Gray wire
Assay Percent Range 99.998% (Metals basis)
Quantity 5 m
Odor Odorless
Form Wire
Chemical Name or Material Lead

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT25I016 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number011901.G1,011901.G5
Lot NumberY20G041 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011901.G1,011901.G5
Lot NumberQ27D026 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number011901.G1,011901.G5
Lot NumberU10H012 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number011901.G1,011901.G5
Lot NumberP19E063 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number011901.G1,011901.G5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.