missing translation for 'onlineSavingsMsg'
Learn More

Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals

Used in piezoelectric devices and electro-optic devices

£84.72 - £1329.00

Chemical Identifiers

CAS 12626-81-2
Molecular Formula O5PbTiZr
Molecular Weight (g/mol) 426.286
MDL Number MFCD00212270
InChI Key HFGPZNIAWCZYJU-UHFFFAOYSA-N
Synonym lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
PubChem CID 159452
IUPAC Name lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334737
View Documents
Thermo Scientific Alfa Aesar
039758.09
10 g
£84.72
10g
Please to purchase this item. Need a web account? Register with us today!
11344737
View Documents
Thermo Scientific Alfa Aesar
039758.18
50 g
£347.73
50g
Please to purchase this item. Need a web account? Register with us today!
11354737
View Documents
Thermo Scientific Alfa Aesar
039758.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12626-81-2
426.286
HFGPZNIAWCZYJU-UHFFFAOYSA-N
159452
[O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
O5PbTiZr
MFCD00212270
lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
lead(2+);oxygen(2-);titanium(4+);zirconium(4+)

Specifications

12626-81-2
Semi-solid, Viscous Liquid
MFCD00212270
lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
HFGPZNIAWCZYJU-UHFFFAOYSA-N
lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
159452
Lead zirconium titanium oxide, Polymeric precursor
Yellow
O5PbTiZr
UN1993
Soluble in alcohols
[O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
426.286
Oxide ≈30 wt%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H304-H373-H318-H302-H336-H315
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes skin irritation.

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P331-P332+P313-P362-P501c

H302+H332-H304-H315-H319-H336-H350-H360Df-H373

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II

missing translation for 'tsca' : Yes

Product Identifier
  • Lead zirconium titanium oxide, polymeric precursor

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Lead zirconium titanium oxide, Toluene,2-Propanol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.