missing translation for 'onlineSavingsMsg'
Learn More
Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals Product Code.: 11354737

Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals

Product Code. 11354737
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11354737

Brand: Thermo Scientific Alfa Aesar 039758.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in piezoelectric devices and electro-optic devices

Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12626-81-2
Molecular Formula O5PbTiZr
Molecular Weight (g/mol) 426.286
MDL Number MFCD00212270
InChI Key HFGPZNIAWCZYJU-UHFFFAOYSA-N
Synonym lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxideShow MoreShow Less
PubChem CID 159452
IUPAC Name lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]

Specifications

Color Yellow
Physical Form Semi-solid, Viscous Liquid
Quantity 250 g
UN Number UN1993
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈30 wt%
Chemical Name or Material Lead zirconium titanium oxide, Polymeric precursor

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM18F064 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog Number039758.09,039758.18,039785.AP,039758.30
Lot NumberL16T016 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039758.09,039758.18,039785.AP,039758.30
Lot NumberI14X002 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039758.09,039758.18,039785.AP,039758.30
Lot NumberC22W035 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039758.09,039758.18,039785.AP,039758.30
Lot NumberD27W006 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039758.09,039758.18,039785.AP,039758.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.