missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Leakproof HDPE Narrow-Mouth Bottles

Economical bottles with a double-seal, leakproof closure.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.83000kg

 View more versions of this product

Product Code. 11735383

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Economical bottles with a double-seal, leakproof closure

  • Translucent HDPE
  • Narrow mouth
  • No drip lip ensures safe, clean pouring
  • Linerless screw caps attached

Specifications

Specifications

1 oz. (30 mL)
1 oz.
HDPE
Polypropylene
Screw Cap
30 mL
Natural
20-410
20-410 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers